Assay Details
| Gene Symbol: PCBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256921.2 | NP_001243850.1 | 1 | 69 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001256921.2 transcripts.
Set Membership: |
|

PCBP1
Genomic Map