Assay Details
| Gene Symbol: RAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256928.1 | NP_001243857.1 | 1 | 1522 | 73 | |
| XM_023633190.1 | XP_023488958.1 | 1730 | 73 | |||
More Information
Set Membership: |
|

RAI2
Genomic Map