Assay Details
| Gene Symbol: KCNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256931.1 | NP_001243860.1 | 1 | 1522 | 77 | |
| XM_023642187.1 | XP_023497955.1 | 2981 | 77 | |||
More Information
Set Membership: |
|

KCNA1
Genomic Map