Assay Details
Gene Symbol: SLITRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257070.1 | NP_001243999.1 | 1 | 2424 | 125 | |
XM_005601178.3 | XP_005601235.1 | 2992 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
