Assay Details
| Gene Symbol: FEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257085.2 | NP_001244014.1 | 1 | 1524 | 93 | |
| XM_005598189.3 | XP_005598246.1 | 1818 | 93 | |||
More Information
Set Membership: |
|

FEN1
Genomic Map