Assay Details
| Gene Symbol: ADRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257120.1 | NP_001244049.1 | 1 | 919 | 71 | |
| GenBank mRNA | JL628735.1 | - | 1 | 83 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADRB2| Gene Symbol: ADRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257120.1 | NP_001244049.1 | 1 | 919 | 71 | |
| GenBank mRNA | JL628735.1 | - | 1 | 83 | 71 | |
Set Membership: |
|