Assay Details
| Gene Symbol: SCG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257121.2 | NP_001244050.2 | 1 | 1720 | 94 | |
| XM_005610603.3 | XP_005610660.2 | 1888 | 94 | |||
More Information
Set Membership: |
|

SCG2
Genomic Map