Assay Details
| Gene Symbol: COA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257140.1 | NP_001244069.1 | 1 - 2 | 124 | 95 | |
| GenBank mRNA | CX594706.1 | - | 1 - 2 | 154 | 95 | |
| CX594789.1 | - | 1 - 2 | 139 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257140.1 transcripts.
Set Membership: |
|

COA4
Genomic Map