Assay Details
| Gene Symbol: CNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257151.1 | NP_001244080.1 | 1 | 2021 | 108 | |
| XM_005596792.3 | XP_005596849.1 | 2690 | 108 | |||
| XM_023649376.1 | XP_023505144.1 | 2773 | 108 | |||
| XM_023649377.1 | XP_023505145.1 | 3105 | 108 | |||
More Information
Set Membership: |
|

CNR1
Genomic Map