Assay Details
| Gene Symbol: ZHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257156.1 | NP_001244085.1 | 1 | 2221 | 66 | |
| XM_023648314.1 | XP_023504082.1 | 2965 | 66 | |||
| XM_023648315.1 | XP_023504083.1 | 2647 | 66 | |||
| XM_023648317.1 | XP_023504085.1 | 2947 | 66 | |||
| XM_023648318.1 | XP_023504086.1 | 2629 | 66 | |||
More Information
Set Membership: |
|

ZHX2
Genomic Map