Assay Details
| Gene Symbol: DKK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267802.1 | NP_001254731.1 | 3 - 4 | 711 | 93 | |
| XM_023642584.1 | XP_023498352.1 | 729 | 93 | |||
| GenBank mRNA | JN966754.1 | - | 3 - 4 | 711 | 93 | |
More Information
Set Membership: |
|

DKK1
Genomic Map