Assay Details
| Gene Symbol: DRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284542.1 | NP_001271471.1 | 4 - 5 | 808 | 88 | |
| GenBank mRNA | JQ254079.1 | - | 4 - 5 | 808 | 88 | |
| JQ254080.1 | - | 4 - 5 | 808 | 88 | ||
| JQ254081.1 | - | 4 - 5 | 808 | 88 | ||
| JQ254082.1 | - | 4 - 5 | 808 | 88 | ||
| JQ254083.1 | - | 4 - 5 | 808 | 88 | ||
More Information
Set Membership: |
|

DRA
Genomic Map