Assay Details
| Gene Symbol: RAP2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301209.2 | NP_001288138.1 | 1 - 2 | 355 | 91 | |
| XM_023633198.1 | XP_023488966.1 | 863 | 91 | |||
| GenBank mRNA | JL616364.1 | - | 1 - 2 | 260 | 91 | |
More Information
Set Membership: |
|

RAP2C
Genomic Map