Assay Details
| Gene Symbol: PRORP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301261.2 | NP_001288190.1 | 4 - 5 | 1700 | 77 | |
| XM_001491047.4 | XP_001491097.1 | 1962 | 77 | |||
| XM_005603440.3 | XP_005603497.1 | 1924 | 77 | |||
| GenBank mRNA | HQ889874.1 | - | 2 - 3 | 170 | 77 | |
| JL624035.1 | - | 3 - 4 | 206 | 77 | ||
More Information
Set Membership: |
|

PRORP
Genomic Map