Assay Details
| Gene Symbol: CNOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301266.2 | NP_001288195.1 | 3 - 4 | 551 | 53 | |
| XM_005610612.3 | XP_005610669.1 | 476 | 53 | |||
More Information
Set Membership: |
|

CNOT9
Genomic Map