Assay Details
| Gene Symbol: DNTTIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309173.2 | NP_001296102.1 | 1 - 2 | 137 | 69 | |
| GenBank mRNA | CD472348.1 | - | 1 - 2 | 63 | 69 | |
| CD528412.1 | - | 1 - 2 | 137 | 69 | ||
More Information
Set Membership: |
|

DNTTIP2
Genomic Map