Assay Details
| Gene Symbol: TIMM17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309300.1 | NP_001296229.1 | 3 - 4 | 202 | 78 | |
| GenBank mRNA | JL619014.1 | - | 3 - 4 | 202 | 78 | |
More Information
Set Membership: |
|

TIMM17A
Genomic Map