Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309313.2 | NP_001296242.1 | 11 - 12 | 2160 | 101 | |
| XM_001502799.5 | XP_001502849.1 | 2258 | 101 | |||
| XM_014734210.2 | XP_014589696.1 | 2303 | 101 | |||
| XM_014734211.2 | XP_014589697.1 | 2267 | 101 | |||
| GenBank mRNA | JL640521.1 | - | 8 - 9 | 1063 | 101 | |
More Information
Set Membership: |
|

MMUT
Genomic Map