Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309313.2 | NP_001296242.1 | 1 - 2 | 430 | 99 | |
| XM_001502799.5 | XP_001502849.1 | 528 | 99 | |||
| XM_014734210.2 | XP_014589696.1 | 573 | 99 | |||
| XM_014734211.2 | XP_014589697.1 | 537 | 99 | |||
| GenBank mRNA | JL626691.1 | - | 1 - 2 | 273 | 99 | |
More Information
Set Membership: |
|

MMUT
Genomic Map