Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309313.2 | NP_001296242.1 | 4 - 5 | 1121 | 80 | |
| XM_001502799.5 | XP_001502849.1 | 1219 | 80 | |||
| XM_014734210.2 | XP_014589696.1 | 1264 | 80 | |||
| XM_014734211.2 | XP_014589697.1 | 1228 | 80 | |||
More Information
Set Membership: |
|

MMUT
Genomic Map