Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309313.2 | NP_001296242.1 | 7 - 8 | 1606 | 71 | |
| XM_001502799.5 | XP_001502849.1 | 1704 | 71 | |||
| XM_014734210.2 | XP_014589696.1 | 1749 | 71 | |||
| XM_014734211.2 | XP_014589697.1 | 1713 | 71 | |||
| GenBank mRNA | HQ890001.1 | - | 2 - 3 | 180 | 71 | |
| JL640521.1 | - | 4 - 5 | 509 | 71 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map