Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309313.2 | NP_001296242.1 | 8 - 9 | 1717 | 104 | |
| XM_001502799.5 | XP_001502849.1 | 1815 | 104 | |||
| XM_014734210.2 | XP_014589696.1 | 1860 | 104 | |||
| XM_014734211.2 | XP_014589697.1 | 1824 | 104 | |||
| GenBank mRNA | HQ890001.1 | - | 3 - 4 | 291 | 104 | |
| JL640521.1 | - | 5 - 6 | 620 | 104 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map