Assay Details
| Gene Symbol: STMN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309408.1 | NP_001296337.1 | 2 - 3 | 100 | 68 | |
| XM_005607328.3 | XP_005607385.1 | 176 | 68 | |||
| GenBank mRNA | JL615867.1 | - | 464 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001309408.1, XM_005607328.3 transcripts.
Set Membership: |
|

STMN1
Genomic Map