Assay Details
| Gene Symbol: STMN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309408.1 | NP_001296337.1 | 4 - 5 | 476 | 74 | |
| XM_005607328.3 | XP_005607385.1 | 552 | 74 | |||
| GenBank mRNA | JL615867.1 | - | 840 | 74 | ||
More Information
Set Membership: |
|

STMN1
Genomic Map