Assay Details
| Gene Symbol: CXCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309480.1 | NP_001296409.1 | 3 - 4 | 388 | 77 | |
| GenBank mRNA | AF053497.1 | - | 2 - 3 | 166 | 77 | |
| CD466627.1 | - | 3 - 4 | 388 | 77 | ||
| CD468235.1 | - | 3 - 4 | 397 | 77 | ||
| CD536402.1 | - | 3 - 4 | 360 | 77 | ||
| CD536724.1 | - | 3 - 4 | 388 | 77 | ||
More Information
Set Membership: |
|

CXCL1
Genomic Map