Assay Details
| Gene Symbol: SLCO1B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317251.1 | NP_001304180.1 | 10 - 11 | 1517 | 85 | |
| XM_005610968.3 | XP_005611025.1 | 1946 | 85 | |||
| XM_023642200.1 | XP_023497968.1 | 1951 | 85 | |||
| XM_023642201.1 | XP_023497969.1 | 1808 | 85 | |||
| GenBank mRNA | LN833581.1 | - | 10 - 11 | 1517 | 85 | |
More Information
Set Membership: |
|

SLCO1B3
Genomic Map