Assay Details
Gene Symbol: LOC100051526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317252.1 | NP_001304181.1 | 4 - 5 | 575 | 98 | |
XM_005609894.3 | XP_005609951.1 | 652 | 98 | |||
XM_005609895.3 | XP_005609952.1 | 675 | 98 | |||
GenBank mRNA | JN795139.1 | - | 4 - 5 | 575 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
