Assay Details
| Gene Symbol: KRT39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346158.1 | NP_001333087.1 | 4 - 5 | 912 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT39| Gene Symbol: KRT39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346158.1 | NP_001333087.1 | 4 - 5 | 912 | 78 | |
Set Membership: |
|