Assay Details
| Gene Symbol: KRT26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346177.1 | NP_001333106.1 | 2 - 3 | 567 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT26| Gene Symbol: KRT26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346177.1 | NP_001333106.1 | 2 - 3 | 567 | 105 | |
Set Membership: |
|