Assay Details
| Gene Symbol: KRT25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346190.1 | NP_001333119.1 | 1 - 2 | 491 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT25| Gene Symbol: KRT25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346190.1 | NP_001333119.1 | 1 - 2 | 491 | 132 | |
Set Membership: |
|