Assay Details
| Gene Symbol: KRT75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346217.1 | NP_001333146.1 | 8 - 9 | 1483 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT75| Gene Symbol: KRT75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346217.1 | NP_001333146.1 | 8 - 9 | 1483 | 67 | |
Set Membership: |
|