Assay Details
| Gene Symbol: CD1E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252304.2 | NP_001239233.1 | 3 - 4 | 731 | 109 | |
| GenBank mRNA | JF749877.1 | - | 3 - 4 | 642 | 109 | |
| JL641477.1 | - | 3 - 4 | 1113 | 109 | ||
More Information
Set Membership: |
|

CD1E2
Genomic Map