Assay Details
| Gene Symbol: CD1E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252304.2 | NP_001239233.1 | 5 - 6 | 1101 | 83 | |
| GenBank mRNA | CD464395.1 | - | 4 - 5 | 83 | ||
| JF749877.1 | - | 5 - 6 | 1012 | 83 | ||
| JL641477.1 | - | 1 - 2 | 743 | 83 | ||
More Information
Set Membership: |
|

CD1E2
Genomic Map