Assay Details
| Gene Symbol: KRT86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346200.1 | NP_001333129.1 | 8 - 9 | 1288 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT86| Gene Symbol: KRT86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346200.1 | NP_001333129.1 | 8 - 9 | 1288 | 110 | |
Set Membership: |
|