Assay Details
| Gene Symbol: RIDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309285.2 | NP_001296214.1 | 1 | 120 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001309285.2 transcripts.
Set Membership: |
|

RIDA
Genomic Map