Assay Details
| Gene Symbol: NEUROD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501210.6 | XP_001501260.1 | 55 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001501210.6 transcripts.
Set Membership: |
|

NEUROD2
Genomic Map