Assay Details
Gene Symbol: CERT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599613.3 | XP_005599670.1 | 1657 | 100 | ||
XM_005599614.3 | XP_005599671.1 | 1657 | 100 | |||
XM_005599616.3 | XP_005599673.1 | 1577 | 100 | |||
XM_005599617.3 | XP_005599674.1 | 1657 | 100 | |||
XM_023618163.1 | XP_023473931.1 | 1577 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
