Assay Details
| Gene Symbol: OMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605108.2 | XP_005605165.1 | 297 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005605108.2 transcripts.
Set Membership: |
|

OMD
Genomic Map