Assay Details
| Gene Symbol: SP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643653.1 | XP_023499421.1 | 6716 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023643653.1 transcripts.
Set Membership: |
|

SP7
Genomic Map