Assay Details
| Gene Symbol: TAFA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005611359.3 | XP_005611416.1 | 1432 | 101 | ||
| XM_023643913.1 | XP_023499681.1 | 1432 | 101 | |||
More Information
Set Membership: |
|

TAFA2
Genomic Map