Assay Details
Gene Symbol: LOC100055155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606631.3 | XP_005606688.1 | 314 | 69 | ||
XM_014736832.2 | XP_014592318.1 | 519 | 69 | |||
XM_014736833.2 | XP_014592319.1 | 216 | 69 | |||
XM_014736834.2 | XP_014592320.1 | 979 | 69 | |||
XM_023631663.1 | XP_023487431.1 | 334 | 69 | |||
XM_023631666.1 | XP_023487434.1 | 334 | 69 | |||
XM_023631667.1 | XP_023487435.1 | 321 | 69 | |||
XM_023631668.1 | XP_023487436.1 | 334 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
