Assay Details
| Gene Symbol: THAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645685.1 | XP_023501453.1 | 264 | 86 | ||
| XM_023645686.1 | XP_023501454.1 | 313 | 86 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

THAP12| Gene Symbol: THAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645685.1 | XP_023501453.1 | 264 | 86 | ||
| XM_023645686.1 | XP_023501454.1 | 313 | 86 | |||
Set Membership: |
|