Assay Details
| Gene Symbol: DENND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504905.5 | XP_001504955.1 | 2890 | 90 | ||
| XM_005612148.3 | XP_005612205.1 | 2711 | 90 | |||
| XM_005612149.3 | XP_005612206.1 | 2808 | 90 | |||
| XM_014741799.2 | XP_014597285.1 | 2952 | 90 | |||
| XM_023646005.1 | XP_023501773.1 | 3041 | 90 | |||
| XM_023646006.1 | XP_023501774.1 | 1622 | 90 | |||

DENND2B
Genomic Map