Assay Details
| Gene Symbol: RPL37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490034.5 | XP_001490084.1 | 360 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL37A| Gene Symbol: RPL37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490034.5 | XP_001490084.1 | 360 | 74 | ||
Set Membership: |
|