Assay Details
| Gene Symbol: LOC100630207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610682.3 | XP_005610739.1 | 2259 | 99 | ||
| XM_005610683.3 | XP_005610740.1 | 2256 | 99 | |||
| XM_005610684.3 | XP_005610741.1 | 2256 | 99 | |||
| XM_005610685.3 | XP_005610742.1 | 2256 | 99 | |||
| XM_005610687.3 | XP_005610744.1 | 2235 | 99 | |||
| XM_005610688.3 | XP_005610745.1 | 2220 | 99 | |||
| XM_005610689.3 | XP_005610746.1 | 2187 | 99 | |||
| XM_005610690.3 | XP_005610747.1 | 2109 | 99 | |||
| XM_005610691.3 | XP_005610748.1 | 2085 | 99 | |||
| XM_023642530.1 | XP_023498298.1 | 2259 | 99 | |||
| XM_023642531.1 | XP_023498299.1 | 2106 | 99 | |||
| XM_023642532.1 | XP_023498300.1 | 2082 | 99 | |||

LOC100630207
Genomic Map