Assay Details
| Gene Symbol: LOC100630207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610682.3 | XP_005610739.1 | 2459 | 104 | ||
| XM_005610683.3 | XP_005610740.1 | 2456 | 104 | |||
| XM_005610684.3 | XP_005610741.1 | 2456 | 104 | |||
| XM_005610685.3 | XP_005610742.1 | 2456 | 104 | |||
| XM_005610687.3 | XP_005610744.1 | 2435 | 104 | |||
| XM_005610688.3 | XP_005610745.1 | 2420 | 104 | |||
| XM_005610689.3 | XP_005610746.1 | 2387 | 104 | |||
| XM_005610690.3 | XP_005610747.1 | 2309 | 104 | |||
| XM_005610691.3 | XP_005610748.1 | 2285 | 104 | |||
| XM_023642530.1 | XP_023498298.1 | 2459 | 104 | |||
| XM_023642531.1 | XP_023498299.1 | 2306 | 104 | |||
| XM_023642532.1 | XP_023498300.1 | 2282 | 104 | |||
| GenBank mRNA | JL639746.1 | - | 1 - 2 | 144 | 104 | |
More Information
Set Membership: |
|

LOC100630207
Genomic Map