Assay Details
Gene Symbol: C4H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498834.4 | XP_001498884.1 | 867 | 75 | ||
XM_023639392.1 | XP_023495160.1 | 810 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
