Assay Details
| Gene Symbol: HOXA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499485.6 | XP_001499535.1 | 3836 | 57 | ||
| XM_023639421.1 | XP_023495189.1 | 820 | 57 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXA7| Gene Symbol: HOXA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499485.6 | XP_001499535.1 | 3836 | 57 | ||
| XM_023639421.1 | XP_023495189.1 | 820 | 57 | |||
Set Membership: |
|