Assay Details
| Gene Symbol: GARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014739135.2 | XP_014594621.2 | 1420 | 64 | ||
| GenBank mRNA | JL616340.2 | - | 8 - 9 | 1002 | 64 | |
More Information
Set Membership: |
|

GARS1
Genomic Map