Assay Details
| Gene Symbol: GPR141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500319.4 | XP_001500369.1 | 262 | 85 | ||
| XM_005609310.3 | XP_005609367.1 | 332 | 85 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001500319.4, XM_005609310.3 transcripts.
Set Membership: |
|

GPR141
Genomic Map